J62630.03
J62630.03EA
67.1
EUR
InStock
J62630.03
Stains-all 95%
Stains-all
Purity: 95%
CAS No.: 7423-31-6
Molecular Formula: C30H27BrN2S2
Formula Weight: 559.58
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Dark gray or green
Beilstein Reference: 3865213
MDL No.: MFCD00078788
Applications: Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids