Synonyms:
(2S,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate
J62740.MA
J62740.MAEA
385
EUR
InStock
J62740.MA
(-)-Gallocatechin gallate
(-)-Gallocatechin gallate
CAS No.: 4233-96-9
Molecular Formula: C22H18O11
Formula Weight: 458.38
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00214298
Applications: An antioxidant from green tea