J62969.03
J62969.03EA
79.2
EUR
InStock
J62969.03
Flunarizine dihydrochloride 99%
Flunarizine dihydrochloride
Purity: 99%
CAS No.: 30484-77-6
Molecular Formula: C26H26F2N2.2HCl
Formula Weight: 477.40
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4284243
MDL No.: MFCD00058198
Applications: A dual sodium/calcium channel blocker; a cerebral and peripheral vasodilator