J63020.MA
J63020.MAEA
234
EUR
InStock
J63020.MA
Calcimycin (A23187) 98%
Calcimycin (A23187)
An antibiotic and calcium ionophore
Purity: 98%
CAS No.: 52665-69-7
Molecular Formula: C29H37N3O6
Formula Weight: 523.62
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in DMSO with warming or sonication. Also soluble in ethanol or ethyl acetate
Beilstein Reference: 1096801
MDL No.: MFCD00151202
White to pale yellow powder
Soluble in DMSO with warming or sonication. Also soluble in ethanol or ethyl acetate