J63922.MB
J63922.MBEA
453
EUR
InStock
J63922.MB
Brompheniramine maleate
Brompheniramine maleate
CAS No.: 980-71-2
Molecular Formula: C16H19BrN2
Formula Weight: 319.24
Physical Form: Powder
Appearance: White
MDL No.: MFCD00865691
Applications: An antihistamine and also an inhibitor of serotonin reuptake