Synonyms:
2,2-Bis(hydroxymethyl)-3-quinuclidinone, 2,2-Bis(hydroxymethyl)quinuclidin-3-one
J64071.MA
J64071.MAEA
153
EUR
InStock
J64071.MA
Prima-1
Prima-1
CAS No.: 5608-24-2
Molecular Formula: C9H15NO3
Formula Weight: 185.20
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD04974196
Applications: A cell-permeable quinuclidinone analog.