Synonyms:
Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
J64329.03
J64329.03EA
108
EUR
InStock
J64329.03
Uridine-5'-diphosphate disodium salt 98%
Uridine-5'-diphosphate disodium salt
Purity: 98%
CAS No.: 27821-45-0
Molecular Formula: C9H12N2Na2O12P2
Formula Weight: 448.13
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to off-white
Beilstein Reference: 8817400
MDL No.: MFCD00149253