Synonyms:
3-(4-Hydroxy-phenyl)-propionic acid-OSu, Water-Soluble Bolton-Hunter Reagent, Sulphosuccinimidyl-3 -(4-hydroxypheynyl) propionate
J64508.03
J64508.03EA
141
EUR
InStock
J64508.03
Bolton-Hunter reagent (sulpho-SHPP)
Bolton-Hunter reagent (sulpho-SHPP)
CAS No.: 34071-95-9
Molecular Formula: C13H13NO5
Formula Weight: 263.25
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 132-133°
Beilstein Reference: 1545011
MDL No.: MFCD00005515
Applications: For iodination of proteins that do not contain tyrosine residues