Synonyms:
1-(3-Ethyl-5-methoxybenzo[d]thiazol-2(3H)-ylidene)propan-2-one
J64690.MF
J64690.MFEA
487
EUR
InStock
J64690.MF
TG003
(E/Z)-TG003
TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
Off-white solid
Soluble in DMSO