Synonyms:
N-Benzyl-p-toluenesulphonaMide, N-Tosylbenzylamine
J64910.MA
J64910.MAEA
195
EUR
InStock
J64910.MA
N-Benzyl-p-toluenesulphonamide
N-Benzyl-p-toluenesulphonamide
CAS No.: 1576-37-0
Molecular Formula: C14H15NO2S
Formula Weight: 261.34
Physical Form: Crystalline powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 115°
MDL No.: MFCD00159328
Applications: A potent inhibitor of calcium-stimulated myosin S1 ATPase