Synonyms:
SB225002, 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea
J65387.MA
J65387.MAEA
236
EUR
InStock
J65387.MA
SB 225002
SB 225002
A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2.
Yellow Solid
Soluble to 100mM in DMSO and to 50mM in ethanol