J65443.06
J65443.06EA
96.4
EUR
InStock
J65443.06
5-Bromo-5-nitro-1,3-dioxane 98%, en poudre
5-Bromo-5-nitro-1,3-dioxane
5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.
- Incompatible with strong oxidising agents
- Storage must be tightly closed in well sealed container
Powder
Attention: Incompatible with strong oxidising agents.Storage must be tightly closed in well sealed container.