Synonyms:
(5E)-5-(4-Ethylbenzylidene)-2-mercapto-1,3-thiazol-4(5H)-one, 5-(4-Ethylbenzylidene)rhodanine, 5-[(4-Ethylphenyl)methylene]-2-thioxo-4-thiazolidinone, 10058-F4
J65650.MA
J65650.MAEA
109
EUR
InStock
J65650.MA
(5E)-5-(4-Ethylbenzylidene)rhodanine
(5E)-5-(4-Ethylbenzylidene)rhodanine
CAS No.: 403811-55-2
Molecular Formula: C12H11NOS2
Formula Weight: 249.35
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Yellow
Solubility: Soluble in DMSO. Soluble in water at concentrations less than 2mg/ml
MDL No.: MFCD00758517
Applications: A c-Myc inhibitor that induces apoptosis