Synonyms:
2-(Carbamoyloxy)-N,N,N-trimethyl-1-propanaminium Chloride
J65919.03
J65919.03EA
45.8
EUR
InStock
J65919.03
Bethanechol chloride 98%
Bethanechol chloride
Purity: 98%
CAS No.: 590-63-6
Molecular Formula: C7H17ClN2O2
Formula Weight: 196.68
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 210-229°
MDL No.: MFCD00055224
Applications: A cholinergic receptor agonist