Synonyms:
Cefsulodin sodium, Sulcephalosporin
J66120.03
J66120.03EA
373
EUR
InStock
J66120.03
Cefsulodin sodium salt hydrate 97%
Cefsulodin sodium salt hydrate
A third generation cephalosporin antibiotic
Purity: 97%
Formula Weight: 554.53(anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 175°
Solubility: Freely soluble in water
MDL No.: MFCD07793338
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.
Freely soluble in water
Attention: Research Use only: Not intended for animal or human diagnostic or therapeutic use.