Synonyms:
Sodium (S)-2-(4-(((2,4-diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioate, Methotrexate disodium salt
J66364.MD
J66364.MDEA
214
EUR
InStock
J66364.MD
Sodium methotrexate 98.5%
Sodium methotrexate
Purity: 98.5%
CAS No.: 7413-34-5
Molecular Formula: C20H20N8Na2O5
Formula Weight: 498.40
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Yellow
Boiling Point: 823°
Solubility: Soluble in water
UN No.: UN2811
MDL No.: MFCD00274560
Applications: Inhibits the metabolism of folic acid