Synonyms:
(2S,3S,5R)-3-((1H-1,2,3-Triazol-1-yl)methyl)-3-methyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Tazobactam acid
J66483.03
J66483.03EA
132
EUR
InStock
J66483.03
Tazobactam ≥94%
Tazobactam
Purity: 94%
CAS No.: 89786-04-9
Molecular Formula: C10H12N4O5S
Formula Weight: 300.29
Storage Temperature: -30°C to -10°C
Flash Point: 110°(230°F)
Physical Form: Powder
Appearance: White to off-white
Melting Point: 115-145°
Boiling Point: 77°
Solubility: Soluble in water
MDL No.: MFCD00867002
Applications: Increases efficacy of beta-lactam antibiotics by inactivating beta-lactamase enzymes of beta-lactam resistant microbes