J66500.03
J66500.03EA
73.9
EUR
InStock
J66500.03
Cloxacillin sodium salt monohydrate ≥95%
Cloxacillin sodium salt monohydrate
Purity: 95%
CAS No.: 7081-44-9
Molecular Formula: C19H17ClN3NaO5S.H2O
Formula Weight: 475.88
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Physical Form: Powder
Appearance: White to off-white
Melting Point: 170°
Boiling Point: 69°
Solubility: Soluble in water
MDL No.: MFCD00150735
Applications: Targets primarily gram positive bacteria, especially -lactamase producing Staphylococci