Synonyms:
4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide
J66565.22
J66565.22EA
219
EUR
InStock
J66565.22
Sulphamethoxazole 98%
Sulphamethoxazole
Purity: 98%
CAS No.: 723-46-6
Molecular Formula: C10H11N3O3S
Formula Weight: 253.28
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to off-white
Melting Point: 168-172°
Boiling Point: 482°
Solubility: Soluble in ethanol or acetone. Very slightly soluble in water
Beilstein Reference: 6732984
MDL No.: MFCD00010546
Applications: A broad spectrum bacteriostatic sulfonamide antibiotic