Raccords haute pression, pour tubes de 360 µm, Nanovolume™
Fournisseur: VICI Jour®
Nanovolume™
JOURJR-C360UPK2EA 288 EUR InStock
JOURJR-C360UPK2 JOURJR-C360PPK JOURJR-C360US66 JOURJR-C360US64 JOURJR-C360US62 JOURJR-C360UFS6 JOURJR-C360CPK JOURJR-C360QUPK6 JOURJR-C360QUPK4 JOURJR-C360UPK4 JOURJR-C360QUPK2 JOURJR-C360UPK6 JOURJR-C360RU.5FS2 JOURJR-C360NFS6 JOURJR-C360RU1S66 JOURJR-C360NFPKG JOURJR-C360RU1PK6 JOURJR-C360UFS2 JOURJR-C360RU.5S66 JOURJR-C360QTPK6 JOURJR-C360UFS4 JOURJR-C360QTPK4 JOURJR-C360RU.5S62 JOURJR-C360C JOURJR-C360RU.5S64 JOURJR-C360NFFS JOURJR-C360ET JOURJR-C360QXPK4 JOURJR-C360QXPK6 JOURJR-C360RU1FS6 JOURJR-C360RU.5FS4 JOURJR-C360QTPK2 JOURJR-C360RU.5FS6 JOURJR-C360QXPK2 JOURJR-C360CFS
Raccords haute pression, pour tubes de 360 µm, Nanovolume™
Raccords et connecteurs
These fittings are designed for 360 µm tubing (metal, fused silica, PEEK or fused silica).
Fittings for metal tubing (<1379 bar) are made of HPLC grade SS, including nut and ferrule, these fittings are optimised for use with stainless or electroformed nickel tubing.
Fittings for fused silica tubing (≥689 bar) are made of HPLC grade SS, with nut and a special ferrule which is precision machined from electroformed nickel, for optimal sealing characteristics, the ferrule is gold plated.
Fittings for PEEK or fused silica tubing ( <689 bar) made of premium grade natural PEEK material. Quick-mount versions have an integral base with double-stick tape to secure fittings to a surface.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité