Browguards with 20 cm polycarbonate visor, Invincible®
Fournisseur: JSP LTD
AFA061-131-100
JSPAAFA061-131-100EA 66.5 EUR InStock
JSPAAFA061-131-100
Browguards with 20 cm polycarbonate visor, Invincible®
Écrans de protection
Faceshield offers protection against medium energy, high-speed particles at 120 m/s (270 mph), liquid droplets, and molten metal.
  • Polycarbonate browguard
  • Aluminium edged polycarbonate visor
  • Wheel ratchet headband
  • Optical class 1
A polycarbonate browguard with nylon wheel ratchet adjustable head harness and aluminium edged clear polycarbonate visor, offering protection against medium energy, high-speed particles at 120 m/s (270 mph), liquid droplets, and molten metal. The optical qualities of the visor comply with the requirements of optical class 1 (highest).
Certifications: Conforms to EN 166.1.B.3.9.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité