Belt bag, Force®8
Fournisseur: JSP LTD
BPT170-011-000
JSPABPT170-011-000EA 322 EUR InStock
JSPABPT170-011-000
Belt bag, Force®8
Sachets Pouches and Belt Cases
Force®8 Belt Bag holds mask and filters when not in use, keeping them free from dirt and damage.
  • Convenient accessibility for Force®8 half-masks
  • Filters and mask remain clean and safe when in bag
  • Increased lifespan for stored items
This robust bag is secured with a wide, comfortable strap and string clip. The main compartment allows for the storage of the Force®8 half-mask without the filters attached. On either side of this main compartment are two more compartments suitable for a filter each. This keeps a complete unit safe and clean.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité