Heating thermostats, JUVO
Fournisseur: JUCHHEIM Laborgerate GmbH
55020.11
JUCH55020.11EA 16400 EUR InStock
JUCH55020.11
Heating thermostats, JUVO
Thermostats à circulation Systèmes à circulation fermée Thermostats chauffants
These thermostats have a special design that ensures trouble-free continuous operation at temperatures up to 350 °C. They are built in stainless steel, with a glandless pump, a visual level gauge and a temperature limiter. If required, an expansion chamber containing a float switch can be fitted.
A double layer of heat transfer fluid in these thermostats suppress the oxidative decomposition of heat transfer fluids over 200 °C.
Marlotherm SH® Dowtherm® or low viscosity silicone oil can be used as heat transfer fluids.
These basic units are fitted with one or two 2 kW heating elements.
Certifications: Complies with safety level 2 of DIN 12879.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité