Beer forcing test refrigerated/heating circulating bath, DYNEO™ DD-1001F-BF
Fournisseur: JULABO GmbH
9021709.D
JULA9021709.DEA 12390 EUR InStock
JULA9021709.D
Beer forcing test refrigerated/heating circulating bath, DYNEO™ DD-1001F-BF
Thermostats à circulation Systèmes à circulation fermée Systèmes de contrôle de température
The JULABO beer forcing test bath in conjunction with a photometer determines the product life of beer before clouding. The simulated aging process is achieved through a programmable temperature profile, which is repeated, until the first clouding develops.
  • Automatic cycles of temperature ramps simulate ageing
  • Pre-programmed temperature profiles for forcing test
  • Program modification possible at any time
  • Large bath opening with insert for 20 bottles, 0,5 litres each (Racks for other bottle sizes on request)
  • Removable Plexiglass® cover
  • RS232 interface for online communication
Units feature a powerful and infinitely adjustable pressure pump, a large colour TFT display, multilingual interface and integrated programmer. The DYNEO DD-1001F-BF has integrated external PT100 connection, and PID3 cascade temperature control.
Max. flow rate / max. pressure: CD: 27 L/min; 0,7 bar
Certifications: Classification III according to DIN 12876-1
Informations de commande: Supplied with stainless steel insert basket for 20×0.5 litre bottles and Plexiglass bath cover.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité