MAGIO™ MX Refrigerated/Heating Circulators with Natural Refrigerant
Fournisseur: JULABO GmbH
9033751.N1 9033752.N1
JULA9033751.N1 JULA9033752.N1
MAGIO™ MX Refrigerated/Heating Circulators with Natural Refrigerant
Thermostats à circulation Systèmes à circulation ouverte Cryothermostats à circulation
The MAGIO™ MX refrigerated/heating circulators operate with a working temperature range of −50 to +200 °C and heating capacity of 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications including reactors to 30 L. An extensive range of accessories is available.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts
  • R1270 natural refrigerant
  • Bright, easy to read touchscreen display
  • Variable speed compressors reduce energy consumption and lower noise
  • Powerful, continuously adjustable pressure/suction pump to 31 L/min with 0,92 bar pressure and 0.34 bar suction
The MAGIO™ user interface with high resolution, modern color TFT touch display has a multi-lingual user interface and shows all important information at a glance. Three large, predefined main screens clearly display data and graphics with various application priorities. All display functions are easy to operate with a fingertip. Programmable with USB port for data-logging and program transfer.
Modern interfaces make remote control easy and practical data management possible as well as allowing integration into process structures. All MAGIO™ units have integrated PT 100 connection, USB interface, RS232/RS285 and Ethernet. Analog interfaces are available as optional accessories (9900100). External PT 100 sensor connection and Stakei port for accessory control.
Certifications: UKCA. DIN 12876-1 Class III.
Informations de commande: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité