CORIO® CD Closed stainless steel bath tanks
Fournisseur: JULABO GmbH
9461701
JULA9461701EA 3080 EUR InStock
JULA9461701
CORIO® CD Closed stainless steel bath tanks
Bains Bains-marie
Open bath circulator with high quality bath tanks made of transparent plastic or stainless steel.
  • Insulated stainless steel tanks
  • Integrated handles in the rim
  • Suitable for use with CORIO C/CD heating immersion circulators
The basic refrigeration baths can be combined with JULABO circulators of the CORIO product series. In combination with these circulators, they are intended for the temperature control of liquid media (bath fluids).
Attention: Danger of burns and property damage if unsuitable bath fluid is used.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité