Solid cabinets, with doors
Fournisseur: JUSTRITE MANUFACTURING
24004
JUSU24004EA 2830 EUR InStock
JUSU24004
Solid cabinets, with doors
Mobilier Armoires de stockage
Sturdy solid wall and chemically resistant polyethylene construction with sealed sump is suitable for highly aggressive liquids.
  • External 2" NPT vent/exhaust connections
  • Padlockable hasps for security against unauthorized use
  • Sturdy adjustable shelf on larger models
  • Leveling feet on all four corners
Shelves on larger units adjust on 76 mm centers to accommodate a wide variety of containers. Sloped slightly to the back, they help direct spills to the rear away from the door and user. Poly hinges, door handle and internal latching mechanism means there are no metal parts to corrode.
Information de livraison: Cabinets include adjustable steel leg levelers and patent pending Haz-Alert™ reflective warning labels in 3 languages.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité