Notrax® 562 Sanitop® Floor Mattings
Fournisseur: JUSTRITE MANUFACTURING
562S0035BL
JUSU562S0035BLEA 52.5 EUR InStock
JUSU562S0035BL
Notrax® 562 Sanitop® Floor Mattings
Tapis de sol Tapis anti-fatigue
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
  • Durable rubber compound
  • Molded alternating large/small hole drainage system keeps moisture and debris free from work area
  • Raised pattern surrounds large holes to channel drainage and provide worker traction
  • Beveled ramps on all 4 sides
  • 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité