Plastic Oily Waste Can
Fournisseur: JUSTRITE MANUFACTURING CO
ECN937FLRD
JUSUECN937FLRDEA 135 EUR InStock
JUSUECN937FLRD
Plastic Oily Waste Can
Bidons de sécurité
Safe collection and storage of oily waste materials, constructed of high density polyethylene (HDPE) and offer fire safe temporary storage of solvent rags, waste rags, shavings and other materials subject to spontaneous combustion.
  • Heavy-wall HDPE construction will not rust or dent and is resistant to most chemical compounds for increased durability
  • Self-closing lid reduces oxygen preventing a fire from starting
  • No seam, leak proof design with molded handles for easy handling
  • Feature self-closing lid with foot lever and with a flared base for added stability
  • Foot pedal provides hands-free opening
  • 'Empty every night' label provides convenient reminder
Ideal for temporary storage of solvent rags, waste rags, shavings and other materials subject to spontaneous combustion - keep one at every workstation.
Certifications: FM approved and meets OSHA and NFPA Code 30 requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité