Whole Blood RNA Purification Mini kit, GeneJET™
Fournisseur: Thermo Fisher Scientific
K0761
K0761EA 338 EUR InStock
K0761
Whole Blood RNA Purification Mini kit, GeneJET™
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
GeneJET™ Whole Blood RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from whole blood and related body fluids. The kit utilises silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 15 minutes. The purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analysis.
  • Efficient: Up to 1,8 µg of total RNA from 500 µl of human blood
  • Convenient: Spin columns are capped and assembled with collection tubes
  • Pure: RNA pure from enzyme inhibitors, proteins, ready to use in all demanding applications; validated in cDNA synthesis and quantitative RT-PCR
  • Fast and simple: Procedure takes less than 15 minutes and does not involve organic extraction or differential RBC lysis
  • Compatible: Suitable for wide variety of samples, including human and animal blood treated with EDTA, citrate or heparin, buffy coat, bone marrow, and related body fluids
Information de livraison: Each kit includes lysis buffer, Wash Buffer 1 (concentrated), Wash Buffer 2 (concentrated), water (nuclease-free), GeneJET™ RNA purification columns (pre-assembled with collection tubes), and collection tubes (1,5 and 2 ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité