Fine Pipette Tips for Long Test Tubes
Fournisseur: KARTELL
0094400
KART0094400EA 1590 EUR InStock
KART0094400
Fine Pipette Tips for Long Test Tubes
Pointes de pipettes
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • High clarity, excellent water-repellent and low retention properties
  • Precise centering of the orifice guarantees reliable directionality of the sample when delivered
  • Able to collect a human biological sample and dispense on growth medium, slide tube/containers to carry out analysis of laboratory diagnosis
To be used whenever it is diffIcult to reach the bottom of test tubes with standard products.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité