Sterile Pipette Filter Tips in Rack
Fournisseur: KARTELL
0963500
KART0963500EA 348 EUR InStock
KART0963500
Sterile Pipette Filter Tips in Rack
Pointes de pipettes
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • Sterilised by gamma radiation
  • Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology
  • Hydrophobic pe filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes
  • Tip orifices are sized to completely empty the tip in the event of over-aspiration
Made of PP and including a filter, these tips are DNase, RNase and DNA free, ATP and metal free, cadmiuim free, apyrogenic and PCR inhibitor free.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité