Skin care dispenser, essential electronic, Scott®
Fournisseur: KIMBERLY CLARK
92147
KIMB92147EA 193 EUR InStock
KIMB92147
Skin care dispenser, essential electronic, Scott®
Distributeurs de savon et distributeurs de savon liquide
The Scott® Essential electronic cassette skincare dispenser system is designed for hand hygiene.
  • It features a long battery life: up 60000 hand washes per 3 D-sized alkaline batteries
  • Easy maintenance with this smart system
  • Low product lights when replacements are needed
Reliable, touchless operation to help reduce cross-contamination and the spread of germs, because users don't have to touch the dispenser to get their soap or sanitiser. The white dispenser automatically dispenses one shot of hand soap or hand sanitiser when a user places their hand beneath the dispenser.
Use this touch-free skin care dispenser with 1,2 L cassettes of Scott® Essential foam soap or hand sanitiser.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité