EPA vials with PP cap, KIMBLE®
Fournisseur: KIMBLEKONTES
66014-303
KIML66014-303EA 308 EUR InStock
KIML66014-303
EPA vials with PP cap, KIMBLE®
Fioles
These EPA water analysis vials with screw thread closures are ideal for the collection and storage of water samples.
  • Open-top
  • Made of borosilicate glass
  • Vials are not pre-cleaned
  • White PP closures with PTFE-faced silicone rubber septa
Certifications: Glass conform to USP Type I and ASTM E438, Type I, Class A requirements.
Conditionnement: Vials are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and are placed into a corrugated shelf-pack.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité