LABO 520 PRO-ACTIVE Laboratory Refrigerator
Fournisseur: KIRSCH PHILIPP
12730
KIRS12730EA 0 EUR InStock
KIRS12730
LABO 520 PRO-ACTIVE Laboratory Refrigerator
Réfrigérateurs
Permanent, proactive monitoring of the performance data
and alerts in the case of deviations. So that you can take
countermeasures in good time before a fault threatens
your chilled goods. Provides world's most accurate temperature
control in refrigerated areas as it has two standard PT1000 sensors.
  • Self-closing door with easy-to-replace plastic magnetic seal frame, lockable
  • Automatic fast defrosting (significantly reduces defrosting periods with minimal temperature variation)
  • Low-noise compressor
  • Power failure alarm
  • Alarms can be forwarded using a potential-free contact (e.g., to a mobile phone with optional GSM-Module or to a control centre)
  • Extra-thick 70 mm energy saving insulation
  • Antifreeze against sub-zero temperatures
  • Ventilation-enforced refrigerating machine
External housing made from galvanised sheet steel (rustproof), with white, anti-scratch powder coating. Interior made from smooth aluminium with colourless protective coating. Gibs (every 15 mm) for flexible interior arrangement.
Extra-thick 70 mm energy saving insulation, made from high-quality, compression-moulded and environmental friendly material.
Door hinge on the right-hand side (by default, see illustration), or the left-hand side, can be retrofitted. Forced-air cooling, switches off automatically when you open the door, ensures a uniform temperature and minimizes temperature deviations.
Key switch protected control panel against tampering. Membrane keyboard with digital temperature display and minimum/maximum temperature memory.
Data documentation can be read via USB interface with KIRSCH datanet software.
Informations de commande: Equipped with castors.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité