Closed cup flash tester
Fournisseur: KOEHLER TECHNISCHE PRODUKTEN
K16592
KOEHK16592EA 12590 EUR InStock
KOEHK16592
Closed cup flash tester
Testeurs de point d'éclair
Rapid tester provides rapid determinations of flash point or sustained burning qualities by using a small sample.
  • One minute test result with a 2 ml sample
  • Convenient semi-automatic operations for flash/no flash tests
Closed cup model specifically used for routine flash point tests from –30 to +300 °C, in quality assurance and environmental compliance testing as well as actual flashpoint for paints, fragrances, hydrocarbons and other liquids.
Certifications: ASTM ; IP 303, ISO 3679, ISO 3680, ISO 9038; DOT CFR 49-173.115; IATA.
Information de livraison: Includes an automatic temperature control with a °C selector switch, a syringe, an electronic timer, an integral NIST traceable thermometer (0 to 300 °C), and an external fuel cylinder valve for connecting to a customer-supplied fuel cylinder or other fuel source.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité