L00675.09
L00675.09EA
47.2
EUR
InStock
L00675.09
Chlorure de benzyltripropylammonium 96%
Chlorure de benzyltripropylammonium
Purity: 96%
CAS No.: 5197-87-5
Molecular Formula: C16H28ClN
Formula Weight: 269.86
Melting Point: ca 190° dec.
Beilstein Reference: 4280671