Synonyms:
2-Methylacetoacetanilide, 2-Acetoacetotoluidide
L02812.36
L02812.36EA
0
EUR
InStock
L02812.36
2'-Méthylacétoacétanilide ≥99%
2'-Méthylacétoacétanilide
Purity: 99+%
CAS No.: 93-68-5
Molecular Formula: C11H13NO2
Formula Weight: 191.23
Flash Point: 143°(289°F)
Odour: Odourless
Beilstein Reference: 2099098
MDL No.: MFCD00008782