Synonyms:
3,4-Diaminobenzohydrazide
L05263.03
L05263.03EA
59.9
EUR
InStock
L05263.03
3,4-Diaminobenzhydrazide 98%
3,4-Diaminobenzhydrazide
Purity: 98%
CAS No.: 103956-09-8
Molecular Formula: C7H10N4O
Formula Weight: 166.18
Melting Point: 148-150°
MDL No.: MFCD00017069