L05334.06
L05334.06EA
101
EUR
InStock
L05334.06
Benzyldimethylsilane 96%
Benzyldimethylsilane
Purity: 96%
CAS No.: 1631-70-5
Molecular Formula: C9H14Si
Formula Weight: 150.30
Flash Point: 53°(127°F)
Refractive Index: 1.5026
Boiling Point: 69-71°/15mm
Beilstein Reference: 2935572
UN No.: UN1993
MDL No.: MFCD00039792