L10151.22
L10151.22EA
110
EUR
InStock
L10151.22
Ethoxytriméthylsilane ≥97%
Ethoxytriméthylsilane
Purity: 97%
CAS No.: 1825-62-3
Molecular Formula: C5H14OSi
Formula Weight: 118.25
Flash Point: -18°(-0°F)
Refractive Index: 1.3740
Odour: Mild
Melting Point: -83°
Boiling Point: 75-76°
Beilstein Reference: 1731950
UN No.: 1993
MDL No.: MFCD00009069