L10957.06
L10957.06EA
182
EUR
InStock
L10957.06
1,2,3,4,5-Pentaméthylcyclopentadiene ≥94%
1,2,3,4,5-Pentaméthylcyclopentadiene
Purity: 94%
CAS No.: 4045-44-7
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 44°(111°F)
Refractive Index: 1.4740
Odour: Light
Boiling Point: 57-58°/13mm
Beilstein Reference: 1849832
UN No.: 3295
MDL No.: MFCD00001354