L11012.03
L11012.03EA
56.5
EUR
InStock
L11012.03
Isothiocyanate de 2-iodophényle ≥97%
Isothiocyanate de 2-iodophényle
Purity: 97%
CAS No.: 98041-44-2
Molecular Formula: C7H4INS
Formula Weight: 261.09
Flash Point: 110°(230°F)
Melting Point: 36-39°
Beilstein Reference: 2716369
UN No.: 2923
MDL No.: MFCD00041057