L12046.04
L12046.04EA
74.3
EUR
InStock
L12046.04
2-(4-Morpholino)ethyl isothiocyanate ≥97%
2-(4-Morpholino)ethyl isothiocyanate
Purity: 97%
CAS No.: 63224-35-1
Molecular Formula: C7H12N2OS
Formula Weight: 172.25
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.5390
Boiling Point: 162-163°/10mm
Beilstein Reference: 1210530
UN No.: 2922
MDL No.: MFCD00041221