L12419.06
L12419.06EA
31.4
EUR
InStock
L12419.06
α,o-Toluenedicarbonitrile tech. 90%, grade Technique
α,o-Toluenedicarbonitrile
Purity: 90%
CAS No.: 3759-28-2
Molecular Formula: C9H6N2
Formula Weight: 142.16
Melting Point: 73-79°
Beilstein Reference: 1940497
UN No.: 3439
MDL No.: MFCD00001895