Synonyms:
2-Fluoro-1,4-dimethoxybenzene
L13505.06
L13505.06EA
88.5
EUR
InStock
L13505.06
1,4-Diméthoxy-2-fluorobenzène ≥97%
1,4-Diméthoxy-2-fluorobenzène
Purity: 97%
CAS No.: 82830-49-7
Molecular Formula: C8H9FO2
Formula Weight: 156.16
Refractive Index: 1.5045
Melting Point: 23-26°
Boiling Point: 119-121°/40mm
Beilstein Reference: 6387735
MDL No.: MFCD00042487