L14000.AP
L14000.AU
L14000.APEA
47.8
EUR
InStock
L14000.AP
L14000.AU
Hydrogène peroxyde 35% (p/p) en solution aqueuse stabilisé
Hydrogène peroxyde
CAS No.: 7722-84-1
Molecular Formula: H2O2
Formula Weight: 34.02
Refractive Index: 1.3350
Odour: Pungent
Melting Point: -26°
Boiling Point: 107°
Beilstein Reference: 3587191
Merck Reference: 14,4798
UN No.: 2014
MDL No.: MFCD00011333