L14523.03
L14523.03EA
109
EUR
InStock
L14523.03
(R)-(-)-2-Methyl-2,4-pentanediol ≥98%
(R)-(-)-2-Methyl-2,4-pentanediol
Purity: 98+%
CAS No.: 99210-90-9
Molecular Formula: C6H14O2
Formula Weight: 118.18
Flash Point: 93°(199°F)
Refractive Index: 1.4275
Boiling Point: 197°
Beilstein Reference: 6051650
Optical Rotation: -21° (neat)
MDL No.: MFCD00064275