L14558.03
L14558.03EA
136
EUR
InStock
L14558.03
2-Amino-6-methyl-4(3H)-quinazolone 96%
2-Amino-6-methyl-4(3H)-quinazolone
Purity: 96%
CAS No.: 50440-82-9
Molecular Formula: C9H9N3O
Formula Weight: 175.19
Melting Point: 300°
Beilstein Reference: 609421
MDL No.: MFCD00236015