Synonyms:
2,2-Dimethylhexanoic acid
L14664.03
L14664.03EA
78.1
EUR
InStock
L14664.03
Acide 2,2-diméthylhexanoïque ≥94%
Acide 2,2-diméthylhexanoïque
Purity: 94%
CAS No.: 813-72-9
Molecular Formula: C8H16O2
Formula Weight: 144.22
Refractive Index: 1.4270
Boiling Point: 216-220°
UN No.: 3265
MDL No.: MFCD00965097